Learn More
Bioassay Systems EnzyChrom™ Superoxide Dismutase Assay Kit. For quantitative determination of superoxide dismutase (SOD) enzyme activity.
SDP
Catalog No. 50489292 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of superoxide dismutase (SOD) enzyme activity and evaluation of drug effects. Key Features: Sensitive and accurate. Linear detection range of 0.05 - 3 U/mL SOD. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD440nm. Samples: Blood, cell, tissue etc. Species: All. Procedure: Assay takes 1 hr. Kit size: 100 tests. Detection limit: 0.05 U/mL.
Catalog No. 50-489-292 Supplier Bioassay Systems Supplier No. ESOD100
For quantitative determination of superoxide dismutase (SOD) enzyme activity and evaluation of drug effects. Key Features: Sensitive and accurate. Linear detection range of 0.05 - 3 U/mL SOD. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD440nm. Samples: Blood, cell, tissue etc. Species: All. Procedure: Assay takes 1 hr. Kit size: 100 tests. Detection limit: 0.05 U/mL.