Learn More
Arbor Assays Superoxide Dismutase (SOD) Colorimetric Activity Kit (2 Plate)
SDP
Supplier:  Arbor Assays K028H1
Encompass_Preferred
The DetectX Superoxide Dismutase (SOD) Activity Kit is designed to quantitatively measure SOD activity in a variety of samples. The assay measures all types of SOD activity, including Cu/Zn, Mn, and FeSOD types.
Catalog No. 50-227-8429
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.