Learn More
Invitrogen™ SuperScript™ IV CellsDirect™ cDNA Synthesis Kit

Description
The SuperScript IV CellsDirect cDNA Synthesis Kit is optimized for the synthesis of first-strand cDNA directly from a mammalian cell lysate without first isolating RNA. Cell lysis and reverse transcription are performed in the same tube, and the resulting first-strand cDNA is ready to use in PCR and qPCR.
Features of the SuperScript IV CellsDirect Synthesis Kit
- Compatible with a wide range of mammalian cell types grown under different treatment conditions
- Single-tube format minimizes reagent loss, sample loss, and handling time
- Total lysate volume can be used in first-strand cDNA synthesis reaction, providing greater yields with a limited number of cells and allowing for detection of rare transcripts
- High-quality cDNA generated for use in a variety of applications, including PCR and qPCR
- Simple protocol takes approximately 37 minutes
- Compatible with TaqMan and SYBR Green qPCR master mixes from various manufacturers
- Optimized performance for 1–10,000 cells per sample, results equivalent to those from purified RNA
Features of the SuperScript IV Reverse Transcriptase included in this kit
- Reduced RNase H activity
- High thermal stability
- High yields of cDNA in the first-strand synthesis reaction for greater sensitivity and enhanced detection of rare transcripts
- Superior tolerance to various RT inhibitors
SuperScript IV reverse transcriptase is a top choice for all RT-PCR and qRT-PCR applications. It is a proprietary MMLV mutant with superior robustness and reliability in RT reactions. It is significantly improved over SuperScript III reverse transcriptase in inhibitor resistance, processivity, and reaction speed, while retaining all the benefits of the previous enzyme, including increased thermostability, highly efficient full-length cDNA synthesis, and reduced RNase activity.
How it works
In traditional RT-PCR, RNA is first isolated from cells in a time-consuming procedure that can lead to a loss of material. Using the SuperScript IV CellsDirect cDNA Synthesis Kit, the cells are lysed and the cDNA is generated from the lysate in a single tube with minimal handling and no sample loss. DNase I treatment is an optional step and can be used to eliminate genomic DNA prior to first-strand synthesis. This kit has been optimized for small cell samples, ranging from 10,000 cells down to a single cell.
Specifications
Specifications
| Content And Storage | • SuperScript IV CellsDirect Lysis Solution (3 x 9 mL) |
| Format | Kit |
| GC-Rich PCR Performance | High |
| Reaction Speed | 10 min. |
| Technique | Reverse Transcription |
| Optimal Reaction Temperature | 50°C |
| Quantity | 500 reactions |
| Reverse Transcriptase | SuperScript IV |
| Ribonuclease H Activity | Reduced |
| Shipping Condition | Dry Ice |
| Show More |
Frequently Asked Questions (FAQs)
The RT master mix contains both random and oligo dT primers.
The Invitrogen ezDNase Enzyme is a novel DNase that is highly specific for double-stranded DNA. It has no activity on single-stranded DNA in RT reactions (primers or probes), or on RNA. The enzyme is also thermolabile—it is inactivated quickly at temperatures typical for the SuperScript IV RT reaction (e.g., 50 degrees C). The additional inactivation step is therefore not required for most RT-qPCR applications. If the RNA sample is to be used for RT-PCR of fragments ≥3 kb, incubate the sample for 5 minutes at 55 degrees C in the presence of 10 mM DTT to inactivate the enzyme.
For RT-qPCR applications we recommend using the Invitrogen SuperScript IV VILO Master Mix. The cDNA synthesis reaction setup with this master mix requires fewer pipetting steps and therefore reduces variation in the data. SuperScript IV RT, as a component of the master mix, offers the highest efficiency of cDNA synthesis step compared to competitors’ products.
The only difference is that the incubation time for the reverse transcription reaction has been reduced to 10 minutes and inactivation time has been reduced to 5 minutes at 85 degrees C.
When compared with SuperScript III RT (and other manufacturers’ RTs) in a synthesis reaction for a 9 kb cDNA, SuperScript IV RT performed successful synthesis in just 10 minutes and did so with comparable (or improved) yield (as shown by gel band density).
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.