Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ IV CellsDirect™ Lysis Reagents

Catalog No. 11750550
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-750-550 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-750-550 Supplier Invitrogen™ Supplier No. 11750550
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The SuperScript IV CellsDirect Lysis Reagents kit includes lysis solution, stop solution, and lysis enhancer. These reagents are part of the SuperScript IV CellsDirect cDNA Synthesis Kit and are made available separately here for use when processing a large number of samples.

The SuperScript IV CellsDirect Lysis Reagents kit includes lysis solution, stop solution, and lysis enhancer. These reagents are part of the SuperScript IV CellsDirect cDNA Synthesis Kit and are made available separately here for use when processing a large number of samples. See the SuperScript IV CellsDirect cDNA Synthesis Kit manual for protocol recommendations.

SuperScript IV CellsDirect Lysis Reagents are optimized for synthesizing first-strand cDNA directly from a mammalian cell lysate without first isolating RNA.

Features of SuperScript IV CellsDirect Lysis Reagents include:

  • Ready-to-use pre-mixed cell lysis solution compatible with a variety of cell-types
  • Optimized performance for 1–10,000 cells per sample; results equivalent to those from purified RNA

Note:
This procedure is optimized for a maximum of 10,000 cells per reaction. If needed, volumes can be adjusted for cell numbers greater than 10,000. Using too many cells per lysis reaction can result in incomplete lysis and/or inhibition of RT-PCR.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• SuperScript IV CellsDirect Lysis Solution (9 mL)
• SuperScript IV CellsDirect Stop Solution (1.25 mL)
• Lysis Enhancer (250 μL)

Store at –20°C.

Format Kit
Technique Reverse Transcription
For Use With SuperScript IV CellsDirect cDNA Synthesis Kit
Quantity 500 reactions
Reverse Transcriptase SuperScript IV
Shipping Condition Dry Ice
No. of Reactions 500 reactions
Reaction Format Master Mix
Reagent Type Reverse Transcription
Starting Material Cells
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.