Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ IV First-Strand Synthesis System

Catalog No. 18091300
Encompass_Preferred
Change view
Click to view available options
Includes:
Kit only
Kit with ezDNase
No. of Reactions:
200 Reactions
50 Reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes No. of Reactions
18-091-300 Kit with ezDNase 200 Reactions
18-091-050 Kit only 50 Reactions
18-091-200 Kit only 200 Reactions
18-091-150 Kit with ezDNase 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-091-300 Supplier Invitrogen™ Supplier No. 18091300
Only null left
Add to Cart
Add to Cart

The SuperScript IV First-Strand Synthesis System for RT-PCR, available with or without ezDNase, is optimized for synthesis of first-strand cDNA from purified poly(A)+ or total RNA.

The SuperScript IV First-Strand Synthesis System for RT-PCR, available with or without ezDNase, is optimized for synthesis of first-strand cDNA from purified poly(A)+ or total RNA. The system includes SuperScript IV Reverse Transcriptase (RT), a proprietary MMLV mutant that produces robust and reliable cDNA synthesis in RT reactions. The SuperScript IV First-Strand Synthesis System with ezDNase has the further ability to remove potential genomic DNA contamination.

The SuperScript IV synthesis system is significantly improved over the SuperScript III-based system in inhibitor resistance, processivity, and reaction speed, while retaining all the benefits of the previous system, including increased thermostability, highly efficient full-length cDNA synthesis, and reduced RNase activity. The SuperScript IV system is designed to provide reliable, consistent, and fast cDNA synthesis in the presence of inhibitors found in a wide variety of samples that cause other currently available RTs to perform inefficiently.

The extremely simplified genomic DNA removal step of the SuperScript IV First-Strand Synthesis System with ezDNase dramatically reduces the time of the entire reverse transcription protocol and reduces possible variation in gene expression due to RNA loss or damage during conventional DNase treatment. The SuperScript IV synthesis system with ezDNase is the top choice for performance and flexibility for RT-PCR applications. We recommend it for all RT-PCR applications, especially when reproducibility and reliability is the primary concern and when inhibitors in the RNA sample may interfere with cDNA synthesis, leading to biases in gene expression studies.

Both SuperScript IV First-Strand Synthesis systems contain all components needed for RT reactions, plus an additional control gene and primers, and provide the flexibility to customize the RT set-up to fit the needs of your application.

Features of the SuperScript IV First-Strand Synthesis System include:
• Significantly improved resistance to a variety of inhibitors that can interfere with cDNA synthesis
• Robust and specific cDNA synthesis in a wide range of sample types
• A faster reverse transcriptase reaction that reduces incubation time from >50 min to 10 min
• Significantly better processivity compared to SuperScript III RT
• Simplified genomic DNA removal step (SuperScript IV synthesis system with ezDNase only)

Enzyme source
Purified from E. coli expressing the pol gene of M-MLV, modified to increase thermostability, half-life, processivity, and inhibitor resistance and to reduce RNase H activity.

Performance and quality testing
Endodeoxyribonuclease, exodeoxyribonuclease, and ribonuclease assays; and yield and length of cDNA product

Unit definition
One unit of SuperScript IV RT is the amount of enzyme required to incorporate 1 nmole of deoxyribonucleotide into acid-precipitable material in 10 min at 37°C using poly(A) oligo(dT)12-18 as a template/primer.

Unit reaction conditions
50 mM Tris-HCl (pH 8.3), 4 mM MgCl2, 10 mM DTT, 50 mM KCl, 0.5 mM dTTP, 0.4 MBq/mL [3H]-dTTP, 0.4 mM poly(A) oligo (dT)12-18 and enzyme in 20 μL for 10 min at 37°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• Oligo(dT)20, 4 x 50 μL (50 μM)
• Random hexamers, 4 x 250 μL (50 ng/μL)
• 5X RT buffer, 4 x 1 mL
• DTT, 4 x 250 μL (0.1 M)
• dNTP mix, 250 μL (10 mM)
• SuperScript IV RT, 4 x 50 μL (10,000 units total at 200 U/μL)
• Ribonuclease Inhibitor, 4 x 100 μL (40 U/μL)
E. coli RNase H, 4 x 50 μL (2 U/μL)
• DEPC-treated water, 4 x 1.2 mL
• Total HeLa RNA, 4 x 20 μL (10 ng/μL)
• Sense Control Primer, 4 x 25 μL (10 μM)
• Antisense Control Primer, 4 x 25 μL (10 μM)
• ezDNase Enzyme, 4 x 50 μL
• 10X ezDNase Buffer, 4 x 100 μL
• Nuclease-free water, 4 x 1.25 mL

Store at -15°C to -25°C.

Format Kit
GC-Rich PCR Performance High
Reaction Speed 10 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C
Quantity Each
Reverse Transcriptase SuperScript IV
Final Product Type First-Strand cDNA
No. of Reactions 200 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 12.3 kb
Starting Material RNA
Includes Kit with ezDNase
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.