Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ IV RT Reaction Buffer

Catalog No. 18090050B
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-090-050B 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-090-050B Supplier Invitrogen™ Supplier No. 18090050B
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

SuperScript IV RT Reaction Buffer is included with, and designed for use with, SuperScript IV Reverse Transcriptase.

SuperScript IV RT Reaction Buffer is included with, and designed for use with, SuperScript IV Reverse Transcriptase. This buffer is optimized to give robust and reliable cDNA synthesis. In cases where additional quantity is required, SuperScript IV RT Reaction Buffer is made available here in a stand-alone format.

For use with

SuperScript IV Reverse Transcriptase (Cat. No 18090010)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
For Use With (Application) Reverse Transcription
Quantity 5 x 1 mL
Includes

• SuperScript IV RT Reaction Buffer (5 x 1 mL)

Store at –20°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.