Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript V Reaction Buffer

Catalog No. 19090010B
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-010B 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-010B Supplier Invitrogen™ Supplier No. 19090010B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen™ SuperScript™ V Reaction Buffer is specifically formulated for use with SuperScript™ V Reverse Transcriptase (RT) to support efficient and reliable first-strand cDNA synthesis.

Invitrogen™ SuperScript™ V Reaction Buffer is specifically formulated for use with SuperScript™ V Reverse Transcriptase (RT) to support efficient and reliable first-strand cDNA synthesis. The buffer is supplied with SuperScript V RT and is also available separately for applications requiring additional reaction buffer.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
For Use With (Application) Reverse Transcription
pH Not Available
Quantity 5 x 1 mL
Includes SuperScript V Reaction Buffer (5 x 1 mL)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.