Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript V VILO Master Mix, Primer-Free, with gDNA Removal

Catalog No. 5001329673
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
1250 Reactions
2000 Reactions
50 Reactions
500 Reactions
Includes:
SuperScript V VILO Master Mix (Primer-Free), No RT Control, Water
SuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water
SuperScript V VILO Master Mix (Primer-free) only
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions Includes
50-013-29673 1250 Reactions SuperScript V VILO Master Mix (Primer-free) only
50-013-29671 50 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, Water
50-013-29672 500 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, Water
11-900-003 2000 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, Water
11-900-111 50 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water
11-900-112 500 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water
11-900-113 2000 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 50-013-29673 Supplier Invitrogen™ Supplier No. 11900004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen™ SuperScript™ V VILO™ Master Mix, Primer-Free is a ready-to-use master mix designed for fast, sensitive, and reproducible cDNA synthesis with the flexibility to select your preferred priming strategy.

Invitrogen™ SuperScript™ V VILO™ Master Mix, Primer-Free is a ready-to-use master mix designed for fast, sensitive, and reproducible cDNA synthesis with the flexibility to select your preferred priming strategy. Powered by SuperScript™ V Reverse Transcriptase, the optimized master mix combines robust performance across a broad range of RNA samples with simplified reaction setup, enhanced reagent stability, and greater workflow flexibility. SuperScript V VILO Master Mix, Primer-Free is also available with ezDNase™ Master Mix for fast and convenient genomic DNA removal prior to reverse transcription.

Features of SuperScript V VILO Master Mix, Primer-Free include:

  • Broad RNA input range—supports sensitive cDNA synthesis from 0.1 pg to 5 μg of total RNA
  • Robust performance with challenging RNA—efficient cDNA synthesis from highly structured, GC-rich, degraded and inhibitor-containing RNA
  • Fast cDNA synthesis—complete reverse transcription in as little as 1-10 minutes
  • Enhanced reagent stability—supports ambient shipping and room-temperature storage for up to 1 month
  • Simple, flexible workflow—ready-to-use format simplifies reaction setup, while room-temperature assembly and extended stability of assembled reactions enable greater workflow flexibility

SuperScript V VILO Master Mix, Primer-Free combines SuperScript V RT, RNase inhibitor, buffer, and dNTPs in a single formulation while allowing users to select and optimize primers for their specific application. This flexibility supports gene-specific primers, oligo(dT), random primers, or customized primer combinations while maintaining the convenience of a ready-to-use master mix.

SuperScript V VILO Master Mix, Primer-Free with gDNA Removal allows streamlined genomic DNA removal step. The dsDNA-specific ezDNase enzyme removes contaminating genomic DNA in as little as 2 minutes without affecting RNA.

SuperScript V VILO Master Mix, Primer-Free also features a DTT-free, animal origin–free formulation for improved sustainability and simplified reagent handling.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage • SuperScript V VILO Master Mix, Primer-Free (5 mL)

Store all contents at:
• -20°C for long-term storage until expiry date
• +5°C for up to 5 months
• Room temperature for up to 1 month
Detection Method qPCR, NGS, PCR
Form Liquid
Format Master Mix
GC-Rich PCR Performance High
PCR Method 2-step RT-qPCR, RT-PCR
Reaction Speed 10 min.
Technique Reverse Transcription
Label or Dye Not Applicable
Optimal Reaction Temperature 50°C
Product Type Reverse Transcription Reagent
Reverse Transcriptase SuperScript V
Ribonuclease H Activity Reduced
Sensitivity 0.1 pg total RNA
Shipping Condition Dry Ice, Wet Ice, Ambient
For Use With (Application) Gene Expression, Real Time PCR (qPCR), RT-PCR, cDNA Synthesis, Next-Generation Sequencing (NGS)
Thermostability Up to 65°
Final Product Type First-Strand cDNA
No. of Reactions 1250 Reactions
Reagent Type Reverse Transcription
Size (Final Product) Up to 20 kb
Starting Material RNA
Includes SuperScript V VILO Master Mix (Primer-free) only
Inhibitor Tolerance (Simple) High
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.