Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript V VILO Master Mix

Catalog No. 11800112
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
1250 Reactions
2000 Reactions
50 Reactions
500 Reactions
Includes:
SuperScript V VILO Master Mix only
SuperScript V VILO Master Mix, No RT Control, Water
SuperScript V VILO Master Mix, No RT Control, ezDNase Master Mix, Water
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions Includes
11-800-112 500 Reactions SuperScript V VILO Master Mix, No RT Control, ezDNase Master Mix, Water
11-800-001 50 Reactions SuperScript V VILO Master Mix, No RT Control, Water
11-800-002 500 Reactions SuperScript V VILO Master Mix, No RT Control, Water
11-800-003 2000 Reactions SuperScript V VILO Master Mix, No RT Control, Water
11-800-111 50 Reactions SuperScript V VILO Master Mix, No RT Control, ezDNase Master Mix, Water
11-800-113 2000 Reactions SuperScript V VILO Master Mix, No RT Control, ezDNase Master Mix, Water
11-800-004 1250 Reactions SuperScript V VILO Master Mix only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 11-800-112 Supplier Invitrogen™ Supplier No. 11800112
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen™ SuperScript™ V VILO™ Master Mix is a ready-to-use master mix designed for fast, sensitive, and reproducible cDNA synthesis in two-step RT-qPCR applications.

Invitrogen™ SuperScript™ V VILO™ Master Mix is a ready-to-use master mix designed for fast, sensitive, and reproducible cDNA synthesis in two-step RT-qPCR applications. Powered by SuperScript™ V Reverse Transcriptase, the optimized master mix combines robust performance across a broad range of RNA samples with simplified reaction setup, enhanced reagent stability, and greater workflow flexibility. SuperScript V VILO Master Mix is also available with ezDNase Master Mix for fast and convenient genomic DNA removal prior to reverse transcription.

Features of SuperScript V VILO Master Mix include:

  • Broad RNA input range—supports sensitive cDNA synthesis from 0.1 pg to 5 μg of total RNA
  • Robust performance with challenging RNA—efficient cDNA synthesis from highly structured, GC-rich, degraded and inhibitor-containing RNA
  • Fast cDNA synthesis—complete reverse transcription in as little as 1-10 minutes
  • Enhanced reagent stability—supports ambient shipping and room-temperature storage for up to 1 month
  • Simple, flexible workflow—ready-to-use format simplifies reaction setup, while room-temperature assembly and extended stability of assembled reactions enable greater workflow flexibility

SuperScript V VILO Master Mix combines SuperScript V RT, RNase inhibitor, buffer, dNTPs, and an optimized blend of oligo(dT) and random primers in a single formulation, simplifying reaction setup and helping reduce variability. Its combination of fast cDNA synthesis, robust performance, and flexible room-temperature handling makes it well suited for routine, high-throughput, and automated RT-qPCR workflows.

SuperScript V VILO Master Mix with gDNA Removal allows streamlined genomic DNA removal step. The dsDNA-specific ezDNase enzyme removes contaminating genomic DNA in as little as 2 minutes without affecting RNA, further simplifying the two-step RT-qPCR workflow.

A DTT-free, animal origin-free formulation combined with reduced cold chain requirements further simplifies reagent handling and supports more sustainable workflows.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage • SuperScript V VILO Master Mix (2 x 1 mL)
• SuperScript V VILO No RT Control (1 mL)
• ezDNase Master Mix (500 μL)
• Nuclease-free water (1.25 mL)

Store all contents at:
• -20°C for long-term storage until expiry date
• +5°C for up to 5 months
• Room temperature for up to 1 month
Detection Method qPCR, NGS, PCR
Form Liquid
Format Master Mix
GC-Rich PCR Performance High
PCR Method 2-step RT-qPCR, RT-PCR
Reaction Speed 10 min.
Technique Reverse Transcription
Label or Dye Not Applicable
Optimal Reaction Temperature 50°C
Product Type Reverse Transcription Reagent
Reverse Transcriptase SuperScript V
Ribonuclease H Activity Reduced
Sensitivity 0.1 pg total RNA
Shipping Condition Dry Ice, Wet Ice, Ambient
For Use With (Application) Gene Expression, Real Time PCR (qPCR), RT-PCR, cDNA Synthesis
Thermostability Up to 65°
Final Product Type First-Strand cDNA
No. of Reactions 500 Reactions
Reagent Type Reverse Transcription
Size (Final Product) Up to 10 kb
Starting Material RNA
Includes SuperScript V VILO Master Mix, No RT Control, ezDNase Master Mix, Water
Inhibitor Tolerance (Simple) High
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.