Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ VILO™ Master Mix

Catalog No. 11755050
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
250 Reactions
50 Reactions
500 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-755-050 50 Reactions
11-755-500 500 Reactions
11-755-250 250 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-755-050 Supplier Invitrogen™ Supplier No. 11755050
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

Includes

(200μL) SuperScript™ III RT, RNaseOUT™ , Recombinant Ribonuclease Inhibitor, a proprietary helper protein, random primers, MgCl2, and dNTPs.

SuperScript VILO Master Mix is a first-strand cDNA synthesis master mix for two-step RT-qPCR applications.

SuperScript VILO Master Mix is a first-strand cDNA synthesis master mix for two-step RT-qPCR applications. The master mix combines SuperScript III Reverse Transcriptase (RT) and other RT reaction components in a single-tube format to enable reliable first-strand synthesis and high cDNA yields from a wide range of input RNA concentrations. The high RT efficiency of SuperScript III RT and the optimized formulation allow the master mix to deliver high linearity in cDNA synthesis across a broad range of input RNA template for two-step quantitative RT-PCR assays.

Features of SuperScript VILO Master Mix include:
• Starting material can range up to 2.5 μg of total RNA in a 20 μL cDNA synthesis reaction
• Recombinant ribonuclease inhibitor safeguards against the degradation of target RNA due to ribonuclease contamination
• RT is engineered with reduced RNase H activity and increased thermal stability

For superior cDNA synthesis performance in RT-qPCR applications
SuperScript IV VILO Master Mix (Cat. No. 11756050) elevates the trusted VILO technology to the next level with the highly processive and thermostable SuperScript IV RT and further optimized buffer. The master mix enables efficient cDNA synthesis at higher temperatures in less time and provides superior cDNA yield and sensitivity even with suboptimal purity or scarce templates.

Order Info

Shipping Condition: Dry Ice.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage

• SuperScript VILO Master Mix (200 μL)

Store at –5 to –30°C.

Detection Method qPCR
Format Master Mix
GC-Rich PCR Performance High
Reaction Speed 60 min.
Technique Reverse Transcription
Optimal Reaction Temperature 42°C
Quantity 50 reactions
Reverse Transcriptase SuperScript III
Ribonuclease H Activity Reduced
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR)
Final Product Type First-Strand cDNA
No. of Reactions 50 Reactions
Reaction Format Master Mix
Reagent Type Reverse Transcription
Size (Final Product) Up to 10 kb
Starting Material RNA
Show More Show Less
What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

What is the difference between SuperScript III RT and the RT in the SuperScript VILO kit?

The SuperScript VILO cDNA Synthesis Kit contains a mix of SuperScript III RT and helper proteins which help to increase the efficiency of the reverse transcription reaction and thus improve yield. The RT in the SuperScript VILO kit is active at 42 degrees C due to the helper proteins.

How much of the first-strand cDNA reaction should I load for PCR?

While the volume is dependent on the starting amount of RNA used for the first-strand synthesis and the abundance of the target gene, we'd recommend starting with 10% of the first-strand reaction for your PCR reaction.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.