Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SuperSignal™ West Femto Maximum Sensitivity Substrate

Catalog No. PI34094 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 mL
100 mL
200 mL
4 x 100 mL
4 x 200 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI34094 20 mL
FER34095X4 4 x 100 mL
PI34095 100 mL
FER34096X4 4 x 200 mL
PI34096 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. PI34094 Supplier Thermo Scientific™ Supplier No. 34094
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultra-sensitive enhanced chemiluminescent (ECL) HRP substrate for low-femtogram-level detection by Western blot analysis.

Thermo Scientific™ SuperSignal West Femto Substrate is an ultra-sensitive enhanced chemiluminescent substrate (ECL substrate) for detection of peroxidase activity from HRP-conjugated antibodies and probes in Western blotting.

SuperSignal West Femto Substrate Maximum Sensitivity Substrate is an ultra-sensitive chemiluminescent substrate system for Western blotting with horseradish peroxidase (HRP) enzyme. The substrate kit enables detection of low femtogram (that's mid-zeptomole!) amounts of protein deposited on nitrocellulose or PVDF membrane and probed with appropriate primary and secondary antibodies. When combined with optimized antibody concentrations and blocking buffers, SuperSignal West Femto Substrate enables detection of target proteins in amounts that are small to be seen with typical ECL substrates.

Highlights:

  • ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes
  • Sensitive – detect low-femtogram (mid-zeptomole) amounts of protein in bands on nitrocellulose or PVDF membranes
  • Quantitative – produces quantitative signal that is measurable over two orders of magnitude
  • Saves on antibody – optimized for very dilute antibody concentrations; dilute primary antibodies 5000- to 100,000-fold and secondary antibodies 100,000 to 500,000-fold (from 1 mg/mL stocks)
  • Intense signal – easy to capture an image by exposure to autoradiography film or via a CCD camera or similar imaging system
  • Excellent signal duration – 8 hours of usable light-output when conditions are optimized
  • Stable reagent – kit components are stable for 1 year at 4°C or 6 months at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Product Type Substrate
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line SuperSignal
Quantity 20 mL
Substrate Horseradish Peroxidase
Recommended Antibody Concentrations 2°: 1:100-500K (2-10ng/mL), 1°: 1:5K (10- 200ng/mL)
Sensitivity Low femtogram-level
Signal Duration 6 to 8 hr.
Sufficient For 2 Mini-Gel Blots, 200 sq. cm of Membrane
Content And Storage Sufficient For: 2 mini-gel blots; 200 cm2 of membrane
• Luminol/Enhancer, 10 mL
• Stable Peroxide Buffer, 10 mL

Store reagents at 4°C.
Show More Show Less
What types of membrane work best with SuperSignal West Femto Maximum Sensitivity Substrate?

Most researchers use nitrocellulose or polyvinyldiflouride (PVDF) membranes with SuperSignal West Femto Maximum Sensitivity Substrate. Both work well, although nitrocellulose seems to be better suited in some applications than PVDF. In addition, charge-modified nylon membrane performs well with this substrate. Please also see our guide for choosing western blot membranes (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/transfer-proteins-western-blot/membranes-transfer-buffers-western-blotting/membranes-western-blotting.html).

Will SuperSignal West Femto Maximum Sensitivity Substrate work with nucleic acid blots (Southern blotting, etc.)?

Yes. However, SuperSignal West Femto Maximum Sensitivity Substrate was optimized for use in Western blot sand is generally not sensitive enough for most nucleic acid protocols. For Southern and Northern blotting, use our North2South Chemiluminescent Nucleic Acid Hybridization and Detection Kit (Cat. No. 17097), North2South Chemiluminescent Substrate for HRP (Cat. No. 17295), or the Chemiluminescent Nucleic Acid Detection Kit, if probing for biotinylated probes (Cat. No. 89880).

I used the SuperSignal West Femto Maximum Sensitivity Substrate. Why is the entire film black or showing a reversed image?

The antibody concentration (primary and secondary) is too high. Use the antibody dilutions described in the product instructions. Most background will disappear when the proper blocking reagent and a HRP conjugate concentration are used.

Why do I see more bands with the SuperSignal West Femto Maximum Sensitivity Substrate than I've seen in the past with other substrates?

Because this substrate is more sensitive than other chemiluminescent substrates you might detect low-abundance proteins that were not visible before. When using a more sensitive substrate, more careful optimization of blocking buffer steps and antibody concentrations is required.

What protocol should I use to strip and reprobe a blot detected with SuperSignal West Femto Maximum Sensitivity Substrate ?

See Tech Tip # 23: Strip and reprobe Western blots.

Can the blots detected with the SuperSignal West Femto Maximum Sensitivity Substrate be reprobed?

Although blots detected with chemiluminescent substrates can be stripped and reprobed, some antigen/antibody systems are sensitive to the stripping procedure and might not yield the same quality of results on a stripped blot compared to a new blot. Only actual experimentation will yield information as to whether a given system will allow reprobing. Please also see Tech Tip # 23: Strip and reprobe Western blots (https://tools.thermofisher.com/content/sfs/brochures/TR0023-strip-reprobe-blots.pdf).

What is the lower detection limit of SuperSignal West Femto Maximum Sensitivity Substrate?

The lower detection limit of SuperSignal West Femto Maximum Sensitivity Substrate is low-femtogram (1 x 10^-15). Please also see Tech Tip # 32 Guide to Enzyme Substrates for Western Blotting (https://tools.thermofisher.com/content/sfs/brochures/TR0032-Substrates-Western.pdf).

Should any reagents be avoided when using the SuperSignal West Femto Maximum Sensitivity Substrate?

Yes. Avoid using sodium azide during and after probing steps involving horseradish peroxidase (HRP), as this will inhibit HRP activity. Sulfide, cyanide, fluoride and superoxide ions also inhibit HRP to some extent.

What is the best blocking reagent to use with SuperSignal West Femto Maximum Sensitivity Substrate?

Background is a relative phenomenon - no blocker will prevent all interactions 100% of the time. While a particular blocking agent may give a perfect signal-to-noise ratio for one set of reaction conditions, it may not work as well for another set of similar conditions. The key is to optimize the system by trying various blocking conditions. Please see Tech Tip # 22 Determine source of non-specific background signal in Western Blots (https://tools.thermofisher.com/content/sfs/brochures/TR0022-Determine-background-cause.pdf).

Is milk a good blocking reagent to use with SuperSignal West Femto Maximum Sensitivity Substrate?

Yes and no. Milk contains variable amounts of biotin so it should not be used with avidin/biotin detection systems. Milk also contains varying amounts of phosphoproteins that may make interfere with anti-phosphotyrosine procedures. A variety of blocking buffers are compatible with the substrate. Please see our Blocking Buffer selection table (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/detect-proteins-western-blot/western-blot-buffers/blocking-buffers-western-blotting.html#table).

How much more sensitive is SuperSignal West Femto Maximum Sensitivity Substrate than the other enhanced chemiluminescent (ECL) substrates?

SuperSignal West Femto Maximum Sensitivity Substrate is about 25- to 50-fold more sensitive than GE Healthcare ECL Substrate or Thermo Scientific ECL Substrate (Cat. No. 32106). SuperSignal West Femto Maximum Sensitivity Substrate is also significantly more sensitive than available chemiluminescent substrates for alkaline phosphatase. Please also see Tech Tip # 32 Guide to Enzyme Substrates for Western Blotting (https://tools.thermofisher.com/content/sfs/brochures/TR0032-Substrates-Western.pdf).

Can I use SuperSignal West Femto Maximum Sensitivity Substrate with the identical protocol (i.e., same dilutions) I used with the SuperSignal West Pico Chemiluminescent Substrate?

No, these SuperSignal substrates require different dilutions. If you follow the SuperSignal West Pico Chemiluminescent Substrate protocol when you use SuperSignal West Femto Maximum Sensitivity Substrate, you will see high background or a decreased signal intensity and or duration. You must use less (more dilute) primary and secondary antibodies with SuperSignal West Femto Maximum Sensitivity Substrate (see product instructions for recommended ranges).

How can I increase the sensitivity of my Western blot?

Consider transferring to a different membrane or using a different detection method. We have observed increased sensitivity when using PVDF membranes in place of nitrocellulose. On PVDF membranes, using as little as 1 µg of total rat brain protein, PKC can be detected with alkaline phosphatase-mediated chromogenic detection in some cases using affinity-purified antibodies at a concentration of 0.5 µg/mL. Detection sensitivity can also be increased by using chemiluminescent detection, especially when using a SuperSignal West enhanced chemiluminescence subtrate (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/detect-proteins-western-blot/western-blot-detection-reagents/detection-technologies-western-blotting/chemiluminescent-western-blot-detection/supersignal-chemiluminescent-substrates.html) such as SuperSignal West Pico PLUS, SuperSignal West Dura, or SuperSignal West Femto. The secondary antibody should be used as recommend by the manufacturer and optimized as needed.

How can I determine if SuperSignal West Femto Maximum Sensitivity Substrate is still functional before using it?

In the darkroom, prepare 2 mL of working substrate solution by mixing 1 mL of luminol enhancer and 1 mL of stable peroxide in a clear test-tube. Add 1 µL of undiluted HRP-conjugate to the working substrate solution. The solution should immediately emit a blue light that will fade over the next several minutes. This glow indicates that the components in the substrate kit are still active/functional. If no light emission is evident, test another source of HRP to determine the root cause.

Can I use SuperSignal West Femto Maximum Sensitivity Substrate with imaging systems?

Yes. The SuperSignal western blot substrates are well suited to most imaging systems including CCD camera units; however, most screen-type phosphorimagers give poor results with the SuperSignal substrates.

Can I "fix" my overexposed film when using SuperSignal West Femto Maximum Sensitivity Substrate?

We offer the Pierce Background Eliminator Kit (Cat. No. 21065) that reduces high background, shading, speckling, and over-exposed bands on exposed film.

How can I increase detection sensitivity with SuperSignal West Femto Maximum Sensitivity Substrate?

Treating the nitrocellulose or PVDF membrane with Western Blot Signal Enhancer (Cat. No. 21050) before blocking can boost the chemiluminescent signal of low-intensity bands.

Can the quality of the membrane affect the detection results when using SuperSignal West Femto Maximum Sensitivity Substrate?

A high-quality membrane can effectively bind more target protein and result in a lower background after detection when using SuperSignal substrates.

Can I strip and reprobe my western blot when using SuperSignal West Femto Maximum Sensitivity Substrate?

Nitrocellulose and PVDF membranes can be stripped following detection with SuperSignal chemiluminescent substrates, for reprobing. Restore Western Blot Stripping Buffer (Cat. No. 21059, 21063) can be used to gently but efficiently remove probes from the western blot membrane without damaging the target protein. In most cases, re-blocking of the membrane is not necessary.

I am using SuperSignal West Femto Maximum Sensitivity Substrate. Why is my signal disappearing after only a few minutes?

The most common cause of premature signal burnout is the use of excessive peroxidase-conjugated probe. During the substrate reaction, an extreme excess of peroxidase will deplete the limited amount of substrate resulting in intense, but short, signal duration. To avoid signal fading, it is imperative to use the peroxidase conjugated probe at a concentration recommended in the SuperSignal West Femto Maximum Sensitivity Substrate manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0011307_SupSig_West_Dura_Extend_Dur_Subs_UG.pdf).

How can I increase the signal intensity when I am using SuperSignal West Femto Maximum Sensitivity Substrate?

The following are possible causes for weak signal intensity:
- Low level of protein transferred from the gel to the membrane. This is particularly true for high molecular weight proteins. Reversible Protein Stain Kits for Nitrocellulose and PVDF membranes (Cat. Nos. 24580 and 24585, respectively) may be used to stain the bands on the membrane to indicate transfer efficiency.
- Over-diluted primary and secondary antibody probes. We suggest using the recommended dilution for the antibody probes for each substrate. More sensitive substrates will require a more dilute antibody solution.
- Short substrate incubation time. For best results, incubate the membrane for 5-15 min in the SuperSignal substrate.

How can I reduce the background signal when I am using SuperSignal West Femto Maximum Sensitivity Substrate?

The following are possible causes for high background signal:
- Insufficient blocking of the membrane
- Using a non-optimized blocker
- Using excess peroxidase-conjugated probe
- Cross-reactivity of the probes to the blocking agent

We recommend testing a variety of blockers to find the best one for a given system.
- Serum-based blockers are a popular choice but have the potential to cross-react with an antibody probe.
- Avoid blocking the membrane with BSA when using an antibody against a bovine source.
- Follow the recommended dilution of the peroxidase-conjugated probe for the chemiluminescent substrate selected to reduce the potential for excessive background.

The following tech tips provide more information that may be useful:
- https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0022-Determine-background-cause.pdf
- https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0024-Optimize-Ab-dilutions.pdf
- https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0032-Substrates-Western.pdf
- https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0067-Chemi-Western-guide.pdf

How is light generated when using the SuperSignal West Femto Maximum Sensitivity Substrate?

SuperSignal West Femto Maximum Sensitivity Substrate is a luminol-based enhanced chemiluminescence (ECL) horseradish peroxidase (HRP) substrate. It is oxidized to an excited state product in the presence of the stable peroxide and horseradish peroxidase. As the product decays to a lower energy state, light is released at 425 nm. The emitted light is then captured on X-ray film or by using a CCD camera.
SuperSignal West Femto Maximum Sensitivity Substrate for HRP is optimized for high sensitivity and long signal duration, making it ideal for cooled CCD camera-based detection systems. Unlike substrates with signals that decline to barely detectable levels in 30-60 min, the signal produced with SuperSignal West Femto Maximum Sensitivity Substrate is stable for 24 hr, allowing multiple film or camera exposures.

When using SuperSignal West Femto Maximum Sensitivity Substrate, what may cause a weak signal or no signal?

The following are potential causes of a weak signal or no signal:
- Proteins did not transfer properly during western blotting
- Insufficient binding to the membrane
- Insufficient amounts of antibodies
- Insufficient amount of antigen
- The blocking buffer is covering the antigen
- Exposure time is too short
The following link contains additional helpful information:
https://assets.thermofisher.com/TFS-Assets/BID/Application-Notes/TR0067-Chemi-Western-guide.pdf

Can I use SuperSignal West Femto Maximum Sensitivity Substrate with the protocol (and antibody dilutions) I have for other SuperSignal substrate formulations?

We do not recommend swapping protocols/antibody dilutions between different SuperSignal substrates. For example, if SuperSignal West Femto Maximum Sensitivity Substrate is used with the protocol for SuperSignal West Femto Maximum Sensitivity Substrate, it would result in high background. We also recommend using antibody dilutions as suggested in the manual for the respective SuperSignal substrate.

When using SuperSignal West Femto Maximum Sensitivity Substrate, how should I reprobe the blot after stripping?

We suggest performing the following steps to reprobe blots after stripping:
1. Wash the blot with wash buffer 2 times for 10 min per wash.
2. Perform the immunoassay starting with the blocking step.

What is the stability and recommended storage condition for SuperSignal West Femto Maximum Sensitivity Substrate?

SuperSignal West Femto Maximum Sensitivity Substrate is stable for one year at 4 degrees C or for six months at room temperature. The recommended storage is at 4 degrees C.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.