Learn More
Supertek™ Graham Condensers
GREENER_CHOICESCI_ED
Catalog No. S09556 Shop All Supertek Scientific Products
Change view
Click to view available options
Length (Metric) Jacket:
300 mm
Catalog No. Length (Metric) Jacket
S09556 300 mm
1 options
Catalog No. S09556 Supplier Supertek™ Supplier No. 16.108.0010
Add to Cart
Add to Cart
Comes in spiral form.
Specifications
Standard Taper Joint No Yes
Length (Metric) Jacket 300 mm
For Use With (Application) Distillation
For Use With (Equipment) Burner
No. of Bulbs 0
Material Borosilicate Glass (3.3)
Type Graham Condenser