Learn More
Cytiva (Formerly Pall Lab) Supor 450 Membrane Disc Filters, 0.45 m - 142 mm, tabbed (25/pkg)
Catalog No. 502063230 Shop All Cytiva Products
Encompass
High Flow Rate Membrane Optimized for Biological, Pharmaceutical, and Sterilizing Filtration Requirements-Fast filtration with superior flow rates and high throughputs -Low protein binding and extensive drug compatibility for critical applications-Saves time and money with fewer filter changes per sample volume-142 and 293 mm discs feature printed tab for instant recognition of pore size and lot number This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3230 Supplier Cytiva Supplier No. 60177
High Flow Rate Membrane Optimized for Biological, Pharmaceutical, and Sterilizing Filtration Requirements-Fast filtration with superior flow rates and high throughputs -Low protein binding and extensive drug compatibility for critical applications-Saves time and money with fewer filter changes per sample volume-142 and 293 mm discs feature printed tab for instant recognition of pore size and lot number This product is not approved for use in medical, clinical, surgical or other patient applications.