Learn More
Cytiva SUPOR DCF CAPSULE .02UM 25PK
Catalog No. 5000330602 Shop All Cytiva Products
Encompass
Supor DCF Capsule Filters 1000 cm 0 2 m presterilized by gamma irradiation 25/pk
Catalog No. 50-003-30602 Supplier Cytiva Supplier No. CFS92SSCCL
Add to Cart
Add to Cart
Request a Quote
Supor DCF Capsule Filters 1000 cm 0 2 m presterilized by gamma irradiation 25/pk