Learn More
ILEX LIFE SCIENCES LLC Supplement Pack for PAN-Biotech Endopan 300 SL Serum-free medium for HUVEC and endothelial cells (9 supplements)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P040065S
Encompass_Preferred
Supplement Pack for Endopan 300 SL Medium. QUANTITY: 9 supplements. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: -20 degrees Celsius. DESCRIPTION: Supplement pack for basal medium Endopan 300 SL. COMPOSITION: Serum substitute Panexin SL-S, EGF (human recombinant Epidermal Growth Factor), FGF-2 (human recombinant basic Fibroblast Growth Factor), VEGF (human recombinant Vascular Endothelial Growth Factor), Vitamin C (Ascorbic Acid phosphate), R3-IGF-1 (human recombinant Insulin-like Growth Factor), GA (Gentamycin/Amphotericin), Hydrocortisone, Heparin. DESCRIPTION: PAN-Biotech Endopan 300 SL is a serum-free medium designed for serum-free culture of endothelial cells directly after isolation. This exclusive product is optimized for the maintenance and expansion of endothelial cells under serum-free culture conditions.
Catalog No. 50-238-8603
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.