Description
- All heating, cooling, and connector assemblies include waist belt and tube with quick-disconnect fitting
- 3M Catalog# V-100 , 3M Catalog# V-300 , 3M Catalog# V-400
Specifications
Specifications
| Description | Vortex Cooling Assembly |
| Product Type | Flow Control Valve |
| For Use With (Application) | Blasting, assembly, mechanical, painting, metal repair, cleaning |
| For Use With (Equipment) | L series loose fitting headgear, whitecap™ helmets and h series hoods, 3M™ belt mounted supplied air respirator |
| Includes | Cooling tube with quick-disconnect fitting, waist belt and holder |
| Model | V-100 |
| Pressure | 60 to 90 psig |