Learn More
Chemglass Life Sciences Support, 3-Leg Tripod, 230mm OAH
Supplier: Chemglass Life Sciences CG3047100
Support, 3-Leg Tripod, 230mm OAH. Tripod has detachable polished legs and a black baked coated iron ring. Overall height is 230mm, ring is 120mm OD x 70mm ID.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.