Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Support Pack Plus

Catalog No. 5018259
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-182-59 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-182-59 Supplier Invitrogen™ Supplier No. BMS414
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

PBS (20x)

Assay Buffer (20x)

Wash Buffer (20x)

Substrate Solution

Stop Solution

plates

10 x 96 tests

Buffer pack for 10 plates (plates included)

Reactivity/Species
Human, Mouse, Rat

Reported Application
ELISA

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Assay Buffer, Solution, Wash Buffer
For Use With (Application) ELISA
Physical Form Liquid
Quantity 10 x 96 Tests
What substrate solution do you recommend using with IL-8 Human Matched Antibody Pair (Cat. No. MS204-3MST)?

We recommend the following formulation when making your own: 1:2 mixture of H2O2 and tetramethylbenzidine.
However we also offer a few alternatives for purchase:
- Pierce TMB Substrate Kit (Cat. No. 34021)
- Support Pack Standard (Cat. No. BMS412) and Support Pack Plus (Cat. No. BMS414). Both of these ELISA Support Packs include stabilized TMB substrate (does not require mixing), assay buffer, wash buffer, stop solution and PBS. Support Pack Plus includes plates.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.