Learn More
Cytiva Polyphenylsulfone Filter Funnel Replacement Support Screen, 25mm (1/pkg)
Catalog No. 502063375 Shop All Cytiva Products
Encompass
The 25 mm filter funnels are ideal for liquid clarification, vacuum filtration, scintillation counting studies, particle contamination analysis, etc.Material options available-The polysulfone filter funnels are less expensive than stainless steel filter funnels and more durable than glass hardwareThe stainless steel filter funnels are corrosion-resitant and offer excellent chemical compatibility.They are available in 50 and 200 mL capacities. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3375 Supplier Cytiva Supplier No. 87265
The 25 mm filter funnels are ideal for liquid clarification, vacuum filtration, scintillation counting studies, particle contamination analysis, etc.Material options available-The polysulfone filter funnels are less expensive than stainless steel filter funnels and more durable than glass hardwareThe stainless steel filter funnels are corrosion-resitant and offer excellent chemical compatibility.They are available in 50 and 200 mL capacities. This product is not approved for use in medical, clinical, surgical or other patient applications.