Learn More
Cytiva (Formerly Pall Lab) Spare Parts and Accessories for 293 mm Disc Filter Holder - Support screen, type 316 stainless steel (1/pkg)
Catalog No. 502063364 Shop All Cytiva Products
Encompass
Convenient Sizes and Durable Materials for Filtration of All Your Laboratory Samples-Design optimizes use of filter area with 15% more EFA than most competitive units, giving higher flow rates and extending the filter's service life.-Sanitary connections are easy to use and suitable for pharmaceutical applications.-Corrosion-resistant type 316 stainless steel construction provides broad chemical resistance. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3364 Supplier Cytiva Supplier No. 72161
Convenient Sizes and Durable Materials for Filtration of All Your Laboratory Samples-Design optimizes use of filter area with 15% more EFA than most competitive units, giving higher flow rates and extending the filter's service life.-Sanitary connections are easy to use and suitable for pharmaceutical applications.-Corrosion-resistant type 316 stainless steel construction provides broad chemical resistance. This product is not approved for use in medical, clinical, surgical or other patient applications.