Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interscience Support Stand
SDP

Support stand

$180.20

Specifications

Type Support Stand
View More Specs

Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
16-948-920
SDP
missing translation for 'viewDocuments'
Interscience
413016
Eppendorf 0.5 mL
Each for $180.20
Only null left
Add to Cart
 
16-948-994
SDP
missing translation for 'viewDocuments'
Interscience
413017
Eppendorf 1.5 mL and 2 mL
Each for $180.20
Only null left
Add to Cart
 
16-948-993
SDP
missing translation for 'viewDocuments'
Interscience
413018
Dilucup
Each for $180.20
Only null left
Add to Cart
 
Description

Description

  • Support stand
  • Available for Eppendorf 0.5 mL, Eppendorf 1.5 and 2 mL, and Dilucup for EasySpiral and EasySpiral Pro
Specifications

Specifications

Support Stand
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.