Learn More
Cytiva (Formerly Pall Lab) Supracap™ 50 depth filter capsule with dual-layer media grade PDE1 (0.1-3 m retention rating) and luer-lock connections
Supplier:  Cytiva SC050PDE1
Encompass
Scalable Capsule with Seitz High Performance Depth Filter Media-Flexible choices in filter media for prefiltration and removal of coarse particulate.-Low hold-up volume allows for increased product recovery and requires low post-use rinse volumes.-For filtration of 3 to 100 L.-Mechanically robust design provides unobstructed process flows, consistent and scalable filtration results, and high filter media integrity. -Completely disposable technology -Encapsulated design reduces operator exposure to potential biohazards. -Tested and certified. Manufactured under a Quality Management System certified to ISO 9000 and ISO 9001, and an Environmental Management System certified to ISO 14001.-Offers flexibility and assurance of application success from development to production scale. Scalable to Palls entire line of traditional SUPRAdisc™ modules, as well as Stax™ capsules. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1847030
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.