Learn More
Bio-Serv™ Supreme Mini-Treats™ Round Nutritional Pellets, Certified

Description
- Appetizing aroma helps to deter rodent maternal cannibalism; chocolate and very berry flavor recommended
- Excellent for training, conditioning or as a reward following a less desirable procedure
- Nutritionally complete 1gm treat that compliments enrichment programs
- Diameter: 11.43mm (0.45 in.)
- Excellent source of vitamin C
- Certified (Contaminant Screened)
- Nutritionally assayed
- Shape: pellet
- Weight: 1gm
For training/conditioning
Certifications
Contaminant Screened
Specifications
Specifications
| Product Type | Nutritional Pellets |
| Calories | 3.46 |
| Components | Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Natural and Artificial Flavors, Vitamin Mix, Calcium Silicate, Magnesium Stearate, L-Cystine, Ascorbic Acid, Food Dye (Very Berry Flavor Only) |
| Flavor | Banana |
| Format | Round |
| For Use With (Application) | Training, conditioning |
| Particle Shape | Pellet |
| Quantity | 1000/bag |
| Size | 1 g |
| Sterility | Non-sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.