Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Supreme Mini-Treats™ Round Nutritional Pellets, Certified
SDP

Catalog No. 14726343
Encompass
Change view
Click to view available options
Flavor:
Banana
Chocolate
Marshmallow
Pina Colada
Very Berry
Quantity:
1000/bag
5000/box
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flavor Quantity
14-726-343 Chocolate 1000/bag
14-726-355 Banana 5000/box
14-726-354 Pina Colada 5000/box
14-726-356 Very Berry 5000/box
14-726-342 Banana 1000/bag
14-726-352 Chocolate 5000/box
14-726-353 Marshmallow 5000/box
14-726-346 Very Berry 1000/bag
14-726-344 Marshmallow 1000/bag
14-726-345 Pina Colada 1000/bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14-726-343 Supplier Bio-Serv™ Supplier No. F054721
Add to Cart
Add to Cart

Nutritional mini-treat that comes in different flavors is a good source of calcium and can be used as a reward for doing a good job

  • Appetizing aroma helps to deter rodent maternal cannibalism; chocolate and very berry flavor recommended
  • Excellent for training, conditioning or as a reward following a less desirable procedure
  • Nutritionally complete 1gm treat that compliments enrichment programs
  • Diameter: 11.43mm (0.45 in.)
  • Excellent source of vitamin C
  • Certified (Contaminant Screened)
  • Nutritionally assayed
  • Shape: pellet
  • Weight: 1gm

For training/conditioning

Certifications

Contaminant Screened

Specifications

Product Type Nutritional Pellets
Calories 3.46
Components Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Natural and Artificial Flavors, Vitamin Mix, Calcium Silicate, Magnesium Stearate, L-Cystine, Ascorbic Acid, Food Dye (Very Berry Flavor Only)
Flavor Chocolate
Format Round
For Use With (Application) Training, conditioning
Particle Shape Pellet
Quantity 1000/bag
Size 1 g
Sterility Non-sterile
Storage Requirements Cool dry place
Diameter (English) 0.45 in.
Diameter (Metric) 0.114 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.