Learn More
Description
- Unique blend of cleaners and skin conditioners help leave skin feeling healthy.
- Non-drying formula.
- Gentle on skin.
- Effective on oil, grease, paint, and adhesives.
Specifications
Specifications
| Product Type | Heavy Duty Hand Cleanser |
| Color | Beige |
| Form | Gritty Liquid |
| For Use With (Application) | Oil, Grease Oil, Based Paint, Adhesives, Mud, Clay, Soil, Brake Dust, Carbon Black, Shellacs, Urethanes, Varnish, Wood Stain, Caulk, Silicone Sealers |
| Quantity | 2/Pk. |
| Size | 2 gal. |
| Volume (English) | 1 gal. |
| Volume (Metric) | 3.78 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
