Learn More
GOJO™ SUPRO MAX™ Hand Cleaner
SDP
Catalog No. 19043867 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
296 mL
1.89 L
3.78 L
Quantity:
2/Pk.
4/Pk.
8/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity
19-043-867 1.89 L 4/Pk.
19-043-868 296 mL 8/Pk.
19-043-866 3.78 L 2/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-043-867 Supplier GOJO™ Supplier No. 097204
Add to Cart
Add to Cart
Hand cleaner is a fast and effective heavy-duty hand cleaner with gentle scrubbers.
  • Unique blend of cleaners and skin conditioners help leave skin feeling healthy.
  • Non-drying formula.
  • Gentle on skin.
  • Effective on oil, grease, paint, and adhesives.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Heavy Duty Hand Cleanser
Color Beige
Form Gritty Liquid
For Use With (Application) Oil, Grease Oil, Based Paint, Adhesives, Mud, Clay, Soil, Brake Dust, Carbon Black, Shellacs, Urethanes, Varnish, Wood Stain, Caulk, Silicone Sealers
Quantity 4/Pk.
Size 0.5 gal.
Volume (English) 0.5 gal.
Volume (Metric) 1.89 L