Learn More
Invitrogen™ SUPT4H1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SPT4H.
Specifications
Specifications
| Antigen | SUPT4H1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SUPT4H1 |
| Gene Accession No. | P63271, P63272 |
| Gene Alias | AL022777; DRB sensitivity-inducing factor 14 kDa subunit; DRB sensitivity-inducing factor small subunit; DRB sensitivity-inducing factor small subunit 1; DSIF p14; DSIF small subunit; DSIF small subunit 1; hSPT4; small hSpt4 subunit; SPT4; SPT4 homolog, DSIF elongation factor subunit; SPT4H; suppressor of Ty 4 homolog 1; suppressor of Ty 4 homolog 1 (S. cerevisiae); suppressor of Ty 4 homolog 2; suppressor of Ty 4A; Supt4a; Supt4h; Supt4h1; Supt4h1a; Supt4h2; transcription elongation factor SPT4; Transcription elongation factor SPT4 1; transcription elongation factor SPT4-A; zgc:92824; zgc:92824 protein |
| Gene Symbols | Supt4a, SUPT4H1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.