Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUPV3L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728279 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8279 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8279 Supplier Proteintech Group Inc Supplier No. 128261AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SUV3, also known as SUPV3L1 (suppressor of var1, 3-like 1), is a 786 amino acid protein that localizes to both the nucleus and the mitochondrial matrix and contains one helicase C-terminal domain, as well as one helicase ATP-binding domain. Expressed in a broad range of tissues, SUV3 interacts with HBXIP and functions as an ATPase DNA/RNA helicase that uses magnesium as a cofactor to catalyze the unwinding of DNA/RNA and RNA/RNA duplexes, thereby playing a role in DNA replication and transcriptional initiation. SUV3 exhibits optimal activity at a pH of 5 and, in addition to its helicase activity, is thought to protect cells from apoptosis and participate in maintaining mitochondrial homeostasis.

Specifications

Antigen SUPV3L1
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SUPV3L1
Gene Accession No. Q5EBA1, Q80YD1, Q8IYB8
Gene Alias SUPV3L1, SUV3, SUV3 like protein 1
Gene Symbols Supv3l1
Host Species Rabbit
Immunogen SUPV3L1 Fusion Protein Ag3542
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294385, 338359, 6832
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.