Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518974
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518974 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518974 Supplier Invitrogen™ Supplier No. PA518974
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MRP subfamily which is involved in multi-drug resistance. This protein functions as a modulator of ATP-sensitive potassium channels and insulin release. Mutations and deficiencies in this protein have been observed in patients with hyperinsulinemic hypoglycemia of infancy, an autosomal recessive disorder of unregulated and high insulin secretion. Mutations have also been associated with non-insulin-dependent diabetes mellitus type II, an autosomal dominant disease of defective insulin secretion. Alternative splicing of this gene has been observed; however, the transcript variants have not been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ABCC8
Gene Accession No. Q09428
Gene Alias ABC36; ABCC8; AM60008PU-N; ATP binding cassette subfamily C member 8; ATP-binding cassette sub-family C member 8; ATP-binding cassette transporter sub-family C member 8; ATP-binding cassette, subfamily C (CFTR/MRP), member 8; ATP-binding cassette, sub-family C (CFTR/MRP), member 8; D930031B21Rik; HHF1; HI; HRINS; LOW QUALITY PROTEIN: ATP-binding cassette sub-family C member 8; MRP8; PH SUR1; PHHI; sulfonylurea receptor; sulfonylurea receptor (hyperinsulinemia); sulfonylurea receptor 1; sulfonylurea receptor subunit 1; sulphonylurea receptor 1; SUR; SUR1; SUR1 protein; SUR1delta2; TNDM2
Gene Symbols ABCC8
Host Species Goat
Immunogen Synthetic peptide sequence (EFDKPEKLLSRKD) corresponding to the C-terminus amino acids of ABCC8 (aa 1558-1571).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6833
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.