Learn More
Justrite™ Sure-Grip™ EX Flammables Safety Can Storage Cabinets With Handles
Shop All Justrite Safety Group Products

Description
- Sure-Grip EX flush-mounted door handle does not interfere with traffic; comes with two keys for security and can be padlocked (padlock not included)
- Exclusive self-latching feature ensures that cabinet doors are safely closed without manually engaging door handle
- Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames
- High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
- Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self-latch engages critical three-point latching system for protection under fire conditions
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
- SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- Meet OSHA and NFPA requirements
Specifications
Specifications
| Dimensions (D x W x H) Interior | 14.56 x 39.5 x 39.62 in. |
| Product Type | Can Storage Cabinet |
| Load Bearing Capacity | 30 gal. |
| Door Style | Self-close |
| Width (Metric) Interior | 100.3 cm |
| Depth (Metric) Exterior | 45.7 cm |
| Height (Metric) Exterior | 112 cm |
| Depth (Metric) Interior | 37 cm |
| Weight (Metric) | 176 kg |
| Height (Metric) Interior | 100.6 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.