Learn More
Description
- All testing materials included in each kit
- Easy to read results
- Can be used with serum or plasma
- Reactive and nonreactive controls with dropper tip vials included
- Storage requirement: 2 to 8°C
- Long shelf life
Specifications
Specifications
| CE Marker | Self-declared |
| Clia Complexity | Moderate |
| Detectable Analytes | Mononucleosis Heterophil Antibodies |
| Detection Method | Latex Agglutination |
| Sample Type | Serum, Plasma |
| Sensitivity | 1:28 Davidsohn Titer |
| Type | Mononucleosis Test |
| Disposable | Single-use |
| Formulation | Suspension of Horse Erythrocytes in Buffer with 0.1% Sodium Azide |
| Includes | Reagent, Controls, Test Cards, Disposable Stirrer Pipets |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
