Description
Available in traditional guaiac format for routine screening applications and extra-sensitive format for more stringent testing
- Take-Home Kits include one FOBT triple slide with on-slide monitors, one mailing envelope, three applicator sticks, tissues and instructions
- Use single slides when bleeding in the gastrointestinal tract is strongly suspected as in persons with symptoms of ulcers, black stools or in post-operative patients
- Use triple slides for serial testing as recommended by the American Cancer Society for screening of asymptomatic patients
- Bottles of developer for use with Fecal Occult Blood Slide Test Systems
- 23-038-033 is for use with normal slides
- 23-038-037 is for use with extra-sensitive slides
For screening purposes only
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Clia Complexity | Waived |
| Type | FOBT Single Slide Test |
| For Use With (Application) | Detection of fecal occult Blood |
| Includes | 100 Single slides with on-slide monitors, 2 x 15mL bottles of developer and 100 applicator sticks |
| Quantity | 100 Pack |