Learn More
Fisher Healthcare™ Sure-Vue™ Fecal Occult Blood Slide Test System
GSA_VASDP
Catalog No. 23038035 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Pack
1000 Pack
20 Pack
80 Pack
Type:
FOBT Developer
FOBT Developer (extra sen)
FOBT Single Slide (extra sen)
FOBT Single Slide Test
FOBT Take Home Kit
FOBT Take Home Kit (extra sen)
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-038035 100 Pack FOBT Single Slide (extra sen)
23-038036 80 Pack FOBT Take Home Kit (extra sen)
23-038031 100 Pack FOBT Single Slide Test
23-038032 80 Pack FOBT Take Home Kit
23-038040 1000 Pack FOBT Single Slide Test
23-038037 20 Pack FOBT Developer (extra sen)
23-038033 20 Pack FOBT Developer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 23-038035
Add to Cart
Add to Cart
This item is not returnable. View return policy
Available in traditional guaiac format for routine screening applications and extra-sensitive format for more stringent testing
  • Take-Home Kits include one FOBT triple slide with on-slide monitors, one mailing envelope, three applicator sticks, tissues and instructions
  • Use single slides when bleeding in the gastrointestinal tract is strongly suspected as in persons with symptoms of ulcers, black stools or in post-operative patients
  • Use triple slides for serial testing as recommended by the American Cancer Society for screening of asymptomatic patients
FOBT Developer
  • Bottles of developer for use with Fecal Occult Blood Slide Test Systems
  • 23-038-033 is for use with normal slides
  • 23-038-037 is for use with extra-sensitive slides
For screening purposes only
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Clia Complexity Waived
Type FOBT Single Slide (extra sen)
For Use With (Application) Detection of fecal occult Blood
Includes 100 Extra sensitive slides, 2 x 15mL bottles of extra sensitive developer and 100 applicator sticks
Quantity 100 Pack