Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SureCast™ Multi-Use Tool, 10-well

Catalog No. HC1010
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HC1010 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HC1010 Supplier Invitrogen™ Supplier No. HC1010
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank

Recommended Storage

Room Temperature

Warranty and Services

Up to 12 months

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) SureCast Vertical Gel System
Gel Size Mini
Product Line SureCast
Type SureCast™ Multi-use Tool, 10-well
Quantity 1 unit
Shipping Condition Room Temperature
Do you offer the SureCast casting reagents separately?

The SureCast Handcast System and reagents are available both as a package or as standalones.

Can I reuse SureCast Glass Plates?

Yes, SureCastGlass Plates can be reused.They are up to 22 times more durable than Bio-Rad glass plates.

Why should I store all SureCast reagents at room temperature

Having everything at room temperature means not having to thaw reagents, thus saving time in gel casting.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.