Learn More
Alkali Scientific ProCycle 0.1mL Biorad CFX/Ther
Supplier: Alkali Scientific PCZ9620
Introducing our 0 1ml Biorad CFX Type 96 Well PCR Plate white (No Skirt) designed to elevate your PCR experiments with a host of exceptional features Universal Compatibility This PCR plate is compatible with a broad range of thermal cyclers ensuring versatility and adaptability in your lab Easy Well Identification High-contrast black alphanumeric indexing facilitates quick and accurate well identification streamlining your workflow Enhanced Fluorescence The white wells are designed to enhance fluorescence reflection enabling precise and sensitive detection of your qPCR samples Optimal Heat Transfer Ultra-thin uniform wells ensure optimal heat transfer promoting high reaction efficiency and reproducibility Sample Protection Well ridges help reduce sample evaporation when sealing with film foil or strip caps preserving the integrity of your samples High Clarity Ultra-Pure Material This PCR plate is manufactured from high-clarity ultra-pure polypropylene meeting FDA 21 CFR 177 1520 a
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.