Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SureLock™ Tandem Midi Blot Module

Catalog No. STM2001
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 module
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STM2001 1 module
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. STM2001 Supplier Invitrogen™ Supplier No. STM2001
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Invitrogen SureLock Tandem Midi Blot Module is a wet transfer device used exclusively with the SureLock Tandem Midi Gel Tank. The tank accommodates one blot module per chamber, or two blot modules total.

The SureLock Tandem Midi Blot Module is compatible with all Invitrogen midi gels (blot size: 8.5 cm x 14 cm). The blot module accommodates one midi gel per wet transfer. The unique design make the module easy to use, while the efficient size of the inner core uses less transfer buffer per transfer (only ∼300 mL per transfer). Since transfer buffer contains methanol, this also results in less hazardous waste requiring special disposal. At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes in 30 minutes and at room temperature.

Features:

  • Integrative design—used with the SureLock Tandem Midi Gel Tank, eliminates the need for a separate transfer tank
  • Efficient size—transfer buffer needed is about one-third of traditional wet-transfer systems reducing methanol/transfer buffer waste and cost
  • Quick set-up (time & materials)
  • Compatible—efficiently transfers all Invitrogen pre-cast midi gels
  • Smart design—anode and cathode halves are hinged and connected by a slider. The design enables all parts to be permanently connected eliminating the need for multiple accessories.
  • Low voltage transfer (25 volts)—highly conductive anode plate separated by only 0.5 inches from the cathode plate enables efficient transfer at 25 volts within 30 minutes
  • Room temperature transfer—eliminates the need to pre-chill buffers overnight and the hassle and messiness of ice baths used with traditional wet-transfer systems that require 100 volts for transfer and generate excessive heat
  • Fast and efficient—30 minute transfer without compromising performance
  • Simple clean up—empty, rinse, and dry (no spills or ice disposal)
  • Optional SureLock Tandem Transfer Tray — can be used to set up wet transfer of midi gels using SureLock Tandem Blot Module
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:
1 SureLock Tandem Midi Blot Module
• 1 Blotting Tweezer
• 1 Blotting Roller
• 4 Blotting Sponges
For Use With (Application) Wet Transfer
For Use With (Equipment) SureLock™ Tandem Midi Gel Tank
Gel Compatibility Midi Precast Protein Gels
Gel Size Midi (8 x 13 cm)
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Wet
Product Line SureLock
Type Blot Module
For Use With SureLock Tandem Midi Gel Tank (STM1001)
Gel Dimensions Midi/Wide Format (8 cm x 13 cm)
Quantity 1 module
Shipping Condition Room Temperature
Warranty 1 year
Dimensions (L x W x H) 8 x 12 x 3
Show More Show Less
Can I use the PowerEase Touch 120W Power Supply to transfer two midi gels with the SureLock Tandem Midi Gel Tank and SureLock Tandem Midi Blot Module?

Unfortunately, the PowerEase Touch 120W power supply has a maximum output of 500 mA and is not suitable for transferring two midi gels. For wet transfer, we recommend using the PowerEase Touch Power Supply 350W or 600W unit, which can deliver up to 3 A. More information about these power supply units can be found on our website.

Can I use the PowerEase Touch 120W Power Supply for transferring gels with the SureLock Tandem Midi Blot Module, Mini Gel Blot Module, or Novex Semi-Dry Blotter?

No, most transfer applications require high current and therefore, we recommend using a high current power supply like PowerEase Touch 350W Power Supply.

Do you offer pre-cut membranes and filters for the SureLock Tandem Midi Blot Module?

We offer the following pre-cut membranes and filters for the SureLock Tandem Midi Blot Module:
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, nitrocellulose, Cat. No. STM2008
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.2 µm, nitrocellulose, Cat. No. STM2007
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, PVDF, Cat. No. STM2006

Which blot module is compatible with the SureLock Tandem Midi Gel Tank?

The SureLock Tandem Midi Blot Module (Cat. No. STM2001) is compatible with the SureLock Tandem Midi Gel Tank.

Can I purchase replacement parts for the SureLock Tandem Midi Blot Module (Cat. No. STM2001)?

Yes, the following replacement parts are available for the SureLock Tandem Midi Blot Module:
- SureLock Tandem Midi Gel Tank Replacement Tank, Cat. No. STM1003
- SureLock Tandem Midi Gel Tank Replacement Lid, Cat. No. STM1002
- SureLock Tandem Midi Gel Tank Replacement Cassette Clamp, Cat. No. STM1004
- Gel Knife, Cat. No. EI9010
- SureLock Tandem Midi Sponge Kit, Cat. No. STM2002
- Blotting Roller, Cat. No. LC2100
- SureLock Tandem Midi Blot Module Replacement Gasket Kit , Cat. No. STM2003
- SureLock Tandem Transfer Tray, Cat. No. STM3001

How many gels can I transfer using the SureLock Tandem Midi Blot Module?

Only one midi gel can be transferred at a time using the SureLock Tandem Midi Blot Module. The SureLock Tandem Mini Gel Tank accommodates one SureLock Tandem Midi Blot Module per chamber, or two blot modules total. Hence, upto two midi gels can be transferred at a time using one SureLock Tandem Mini Gel Tank fitted with two SureLock Tandem Midi Blot Modules.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.