Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SureLock™ Tandem Midi Gel Tank Replacement Cassette Clamp

Catalog No. STM1004
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 clamp
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STM1004 1 clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. STM1004 Supplier Invitrogen™ Supplier No. STM1004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This cassette clamp is a replacement part for the SureLock Tandem Midi Gel Tank

The clamp locks and seals the gel cassette within the Gel Runner Tank. The clamp consists of a cam plate and two independently moving cam handles.

About the SureLock Tandem Midi Gel Tank

The SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a set-up time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) SureLock Tandem Midi Gel Tank
Gel Size Midi
Product Line SureLock
Type Midi Gel Tank Replacement Cassette Clamp
Quantity 1 clamp
Shipping Condition Room Temperature
Warranty 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.