Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SureLock™ Tandem Transfer Tray

Catalog No. STM3001
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Tray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STM3001 1 Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. STM3001 Supplier Invitrogen™ Supplier No. STM3001
  • $114.00
    $110.65 / Each of 1

    Save $3.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Invitrogen SureLock Tandem Transfer Tray is used to facilitate wet transfer of midi gels using the SureLock Tandem Blot Module and SureLock Tandem Midi Gel Tank.

The tray has two compartments: the front compartment is used to soak sponge pads and filter papers, and the back compartment is used to position the SureLock Tandem Blot Module to build a transfer stack. The tray has graphics showing the correct orientation for stack assembly and two pour spouts for spill proof pouring of transfer buffer.

About the SureLock Tandem Midi Blot Module

The SureLock Tandem Midi Blot Module is a wet transfer device used exclusively with the SureLock Tandem Midi Gel Tank. The tank accommodates one blot module per chamber, or two blot modules total. The SureLock Tandem Midi Blot Module is compatible with all Invitrogen midi gels (blot size: 8.5 cm x 14 cm). The blot module accommodates one midi gel per wet transfer. The unique design make the module easy to use, while the efficient size of the inner core uses less transfer buffer per transfer (only ∼300 mL per transfer). Since transfer buffer contains methanol, this also results in less hazardous waste requiring special disposal. At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes in 30 minutes and at room temperature.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Wet Transfer
For Use With (Equipment) SureLock™ Tandem Midi Blot Module
Mode of Transfer Wet
Product Line SureLock
Type Transfer Tray
For Use With SureLock Tandem Midi Blot Module (STM2001)
Quantity 1 Tray
Shipping Condition Room Temperature
Warranty 1 year
Dimensions (L x W x H) 14 in. x 10 in. x 3 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.