Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XCell SureLock™ Mini-Cell and XCell II™ Blot Module

Catalog No. ei0002
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EI0002 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EI0002 Supplier Invitrogen™ Supplier No. EI0002
Only null left
Add to Cart
Add to Cart

Includes

XCell II Blot Module; Gel Tension Wedge; Buffer Dam; Lower Buffer Chamber; Buffer Core; Mini-Cell Lid; Gel Knife

Quickly and easily run and blot Novex minigels

The XCell SureLock Mini-Cell and XCell II Blot Module includes the XCell SureLock Mini-Cell for running Novex minigels and the XCell II Blot Module for blotting Novex minigels. The XCell SureLock Mini-Cell allows you to run Novex minigels quickly, easily, and leak-free without any clamps or grease. The incorporation of a gel tension wedge, in place of the rear wedge used on earlier models, makes the XCell SureLock Mini-Cell easy-to-use.

How the system works

  • The XCell SureLock Mini-Cell holds gels firmly with a simple gel tension wedge
  • When the lever on the gel tension wedge is pushed forward into the locked position, an even, horizontal force is generated, sealing the gel/buffer core assembly firmly into position in the lower buffer chamber
  • The positive locking action of the gel tension wedge ensures a trouble-free, leak-free gel run every time
Easy set-up procedure
  • The XCell SureLock Mini-Cell can be easily set up in just three simple steps
  • The XCell SureLock Mini-Cell is available separately or as a part of a complete package
The XCell II Blot Module
  • Allows easy transfer of proteins or nucleic acids from minigels to membranes
  • Fits neatly into the XCell SureLock and XCell II Mini-Cells in place of the gel/buffer core assembly and requires less than 200mL of transfer buffer for Western, Southern, and Northern transfers
  • Tough platinized titanium and stainless steel electrodes create a uniform electrical field without clamps or hinged gel holders
  • Maximum blot size is 9 x 9cm

Upgrade XCell II to XCell SureLock Mini-Cell

  • Conversion of an XCell II into an XCell SureLock is easy with the SureLock Retrofit Kit (available separately)
  • Kit consists of a SureLock Mini-Cell lid, gel tension wedge, and molded minigel buffer dam

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:
• XCell II™ Blot Module
• 1 Gel Tension Wedge
• 1 Buffer Dam
• 1 Lower Buffer Chamber
• 1 Buffer Core
• 1 Mini-Cell Lid
• 1 Gel Knife

Warrantied for one year from the time of purchasing.
Capacity Mini Precast Protein Gels
For Use With (Application) Wet Transfer
For Use With (Equipment) XCell SureLock™ Mini-Cell
Gel Compatibility Mini Precast Protein Gels
Gel Size Mini (8 x 8 cm)
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Wet
Product Line Novex, XCell II, XCell SureLock
Type Blot Module
Quantity 1 unit
Running Dimension Vertical
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.