Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SureSTART™ Cap AVCS™ Screw, 9 mm
GSA_VA

Catalog No. 03378625 Shop All Thermo Scientific Products
Change view
Click to view available options
Closure Size:
9 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
03-378-625 9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-378-625 Supplier Thermo Scientific™ Supplier No. 6ESC950
Get 1 Extra Pack of Vials When You Buy 2
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Screw cap with integral polypropylene septum and thinned penetration area.

  • Screw Cap
  • 9 mm
  • Clear
  • Polypropylene with integral polypropylene septum
  • Thinned penetration Area
  • 100 per pack

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Includes Cap Only
Pre-sterilized No
Liner Material Unlined
Product Type Screw Cap
Closure Size 9 mm
Autoclavable Not Autoclavable
Material Polypropylene
Color Clear
Sterility Non-sterile
For Use With (Equipment) Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.