Learn More
Description
Quickly and accurately detect microorganisms in a broad range of food and environmental samples using the Thermo Scientific™ SureTect™ System. The system combines a real-time PCR instrument with bespoke software and convenient pre-filled reagent components that, together with a single PCR protocol for a range of common targets and sample types, streamline the test workflow.
- Fast Time to Result: Takes less than 24 hours from sample enrichment to results
- Single Enrichment Step: Requires no secondary enrichment or regrowth, for faster, simpler testing
- Easy-to-Use Assay: Requires few pipetting steps, pre-filled lysis tubes and freeze-dried PCR pellets, with no centrifuge or vortex required for sample preparation
- Streamlined Protocol: Facilitates processing of multiple assays in the same run, for maximum efficiency
- Superior PCR Technology: Uses probe-based real-time assays for unparalleled sensitivity and specificity
- Simple, Intuitive Software: Enables rapid, straightforward PCR set-up, automatic data interpretation and traceability of results
- Small Instrument Footprint: Fits easily into laboratory environments, where bench space is at a premium
Specifications
Specifications
| Content And Storage | SureTect Software |
| For Use With (Equipment) | Real Time PCR System |
| For Use With (Application) | SureTect Real Time PCR Assays |
| Description | SureTect Software |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.