Learn More
3M™ Clean-Trace™ Surface Protein (Allergen) Test Swab ALLTEC60, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S27255 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
60/Cs.
Catalog No. Quantity
S27255 60/Cs.
1 options
Catalog No. S27255 Supplier 3M™ Clean-Trace™ Supplier No. ALLTEC60
Add to Cart
Add to Cart
Rapid, simple, reliable solution to monitor the presence of allergens on surfaces to effectively manage cleaning
  • Detects as little as 3 μg of protein on surfaces and in hard to reach areas
  • Provides a tool to help assess and improve hygienic performance
  • Visual reading of color change indicates level of cleanliness
  • Real time results allow immediate corrective action
  • Validated for a range of allergenic proteins including egg, milk, gluten, soy, peanut, almond and buckwheat
Specifications
Detects Allergenic proteins including egg, milk, gluten, soy, peanut, almond and buckwheat
Product Type Protein Test Swab
Test Type Surface Hygiene
For Use With (Application) Monitoring allergens on surfaces
Quantity 60/Cs.