Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Surface Temperature Sensor
SDPSCI_ED

Catalog No. S16259ND Shop All Vernier Software Products
Change view
Click to view available options
Temperature Range (Metric):
-25°C to +125°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
S16259ND -25°C to +125°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16259ND Supplier Vernier™ Supplier No. STSBTA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Vernier Surface Temperature Sensor

Requires

Requires an interface and software, sold separately

The Surface Temperature Sensor is designed for use in situations in which low thermal mass or flexibility is required, or for a skin temperature measurement.

Temperature range: –25 to 125°C (–13 to 257°F) Maximum temperature that the sensor can tolerate without damage: 150°C Typical Resolution: 0.08°C (–25 to 0°C) 0.03°C (0 to 40°C) 0.1°C (40 to 100°C) 0.25°C (100 to 125°C) Temperature sensor: 20 kΩ NTC Thermistor Accuracy: ±0.2°C at 0°C, ±0.5°C at 100°C Response time (time for 90% change in reading) 50 seconds (in still air) 20 seconds (in moving air) Probe dimensions: Probe length (handle plus body) 15.5 cm Important: For use in air and water only. For temperature measurements in harsher environments, we recommend the Stainless Steel Temperature Sensor.

Warranty and Services

5-year limited warranty

Specifications

Grade K-8, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Accuracy ±0.2°C at 0°C, ±0.5°C at 100°C
Resolution 0.08°C (-25 to 0°C), 0.03°C (0 to 40°C), 0.1°C (40 to 100°C), 0.25°C (100 to 125°C)
Response Time 50 seconds (in still air), 20 seconds (in moving air)
Temperature Range (Metric) -25°C to +125°C
Length (Metric) 15.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.