Learn More
Cytiva Surfactant P20, Supplied as a Sterile, Filtered 10% Solution in Water
Catalog No. 501055279 Shop All Cytiva Products
Encompass
Buffer additive reducing adsorption of hydrophilic molecules to flow system surfaces.
Catalog No. 50-105-5279 Supplier Cytiva Supplier No. BR100054
Buffer additive reducing adsorption of hydrophilic molecules to flow system surfaces.