Learn More
Description
- Terumo's soft, flexible interlocking wings readily conform to the body's contour
- The wing is angled for ease of positioning of the needle during venipuncture, or less risk of vessel damage or occlusion of the lumen
- The precisely honed needle with thin wall design assures ease of penetration and continued access with a single insertion
Specifications
Specifications
| Product Type | Winged Infusion Set |
| Color | Blue |
| Material | PVC Tubing |
| Needle Gauge | 23 G |
| Sterility | Sterile |
| Latex Free | Yes |
| Length (English) Needle | 0.75 in. |
| Length (English) Tube | 3.5 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.