Learn More
Essendant Protect It! Surge Protector, 7 AC Outlets, 7 ft Cord, 2,160 J, Black
Supplier:  Essendant TRPSUPER7B
Encompass_Preferred
Protect workstations, desktop PCs and other sensitive electronics from surges and line noise with this surge suppressor. Transformer-friendly design accommodates at least one large transformer plug without covering remaining outlets. Features Diagnostic LEDs to confirm outlet grounding and surge suppression status.
Catalog No. 26-779-89
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.