Learn More
Poltex Surgeon's Cap Holder
SDP
Catalog No. 22000466
Change view
Click to view available options
Mounting:
Magnetic Mount
Wall Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-466 Wall Mount
22-000-465 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Consumables shown for demonstration purposes only and are not included with the product.
Catalog No. 22-000-466 Supplier Poltex Supplier No. SURGCAPW
Add to Cart
Add to Cart
Consumables shown for demonstration purposes only and are not included with the product.
Holds and dispenses one box of surgeon's caps.
  • Holds and dispenses one box of surgeons caps
  • Super durable and cleanable (alcohol resistant) PETG construction.
  • Choose from mounting methods:
    • Powerful Magnets
    • Keyhole slots (note: mounting hardware not included. Customer must provide their own screw/anchor for their wall type)
  • Fits Medline NON28626 or equivalent box.
  • See image for dimensions
Specifications
Product Type Surgeon's Cap Holder
Material PETG
Mounting Wall Mount
For Use With (Application) Healthcare
For Use With (Equipment) Surgeon's cap