Learn More
Surgical Design No. 60 Carbon Scalpel Blade
Surgical Design™ Royaltek™ Post Mortem Autopsy Scalpel Blades are made from surgical carbon steel blades and sharpened to perfection for a super clean cut.
Supplier:  Surgical Design BLADE#60
Catalog No. 22-079-682
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Surgical Design™ presents Royaltek™- our premium line of surgical blades
  • Convex cutting edge
  • Thicker blades reduce breakage
  • Exceptionally strong and unifoirmly sharpened to maximum acuity
  • Surgical-quality blades
  • Individually sealed
  • Peelable packing
  • Sterile
  • Fits no.8 scalpel handle 
Specifications
Specifications
Carbon Steel
Scalpel Blade
# 60
Single-use
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.