Learn More
Surgical Design Safety Scalpels
Catalog No. 22079718 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 10
# 11
# 12
# 15
# 20
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-079-718 # 12
22-079-716 # 10
22-079-719 # 15
22-079-720 # 20
22-079-717 # 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-079-718 Supplier Surgical Design Supplier No. SSCALPEL#12
Add to Cart
Add to Cart
This item is not returnable. View return policy
Top activated retractable safety scalpel
  • Audible clicks indicate blade position (retracted, extended and final lock for disposal)
  • With non-slip ribbed handle
Specifications
Product Type Safety Scalpel
Size # 12