Learn More
Stoelting™ Surgical Drapes
Stoelting™ Surgical Drapes are for use in minor surgical or therapeutic procedures where a sterile field or drape is required.
$414.00 - $426.00
Specifications
For Use With (Application) | Neuroscience |
---|---|
Dimensions (L x W) | 18 x 26 in. |
Includes | Case of 300 |
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
10-000-695
![]() |
Stoelting™
50980 |
Fenestrated |
Case of 300 for $426.00
|
|
|||||
10-000-696
![]() |
Stoelting™
50981 |
Polyback |
Case of 300 for $414.00
|
|
|||||
Description
- Poly interlined
- 3-ply construction of tissue/poly/tissue provides superior absorbency
- Avoids possible moisture or bacteria migration.
- Available plain or fenestrated
- Sterile
Specifications
Neuroscience | |
18 x 26 in. | |
Case of 300 |